missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetyl-2,5-dimethylfuran, 98%

Manufacturer:  Alfa Aesar™ A1554530

 View more versions of this product

Catalog No. AAA1554530



Specifications

Specifications

3-Acetyl-2,5-dimethylfuran
198°C
10599-70-9
MFCD00010281
3-acetyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl ethanone, ethanone, 1-2,5-dimethyl-3-furanyl, 3-acetyl-2,5-dimethyl furan, 2,5-dimethyl-3-acetylfuran, 1-2,5-dimethyl-3-furyl ethan-1-one, 2,5-dimethyl-3-furyl methyl ketone, 1-2,5-dimethyl-3-furanyl ethanone, 1-2,5-dimethyl-3-furyl ethanone, unii-798v2t7zbv
CC1=CC(=C(O1)C)C(=O)C
138.166
138.17
78°C (172°F)
250g
C8H10O2
112094
KBSVBCHYXYXDAG-UHFFFAOYSA-N
1-(2,5-dimethylfuran-3-yl)ethanone
61527
98%

Chemical Identifiers

10599-70-9
138.166
KBSVBCHYXYXDAG-UHFFFAOYSA-N
61527
CC1=CC(=C(O1)C)C(=O)C
C8H10O2
MFCD00010281
3-acetyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl ethanone, ethanone, 1-2,5-dimethyl-3-furanyl, 3-acetyl-2,5-dimethyl furan, 2,5-dimethyl-3-acetylfuran, 1-2,5-dimethyl-3-furyl ethan-1-one, 2,5-dimethyl-3-furyl methyl ketone, 1-2,5-dimethyl-3-furanyl ethanone, 1-2,5-dimethyl-3-furyl ethanone, unii-798v2t7zbv
1-(2,5-dimethylfuran-3-yl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H319-H317
Toxic if swallowed.
Combustible liquid.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents