Learn More
Alfa Aesar™ 3-Acetyl-2,5-dimethylfuran, 98%
Manufacturer: Alfa Aesar™ A1554518
Specifications
3-Acetyl-2,5-dimethylfuran | |
198°C | |
10599-70-9 | |
MFCD00010281 | |
3-acetyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl ethanone, ethanone, 1-2,5-dimethyl-3-furanyl, 3-acetyl-2,5-dimethyl furan, 2,5-dimethyl-3-acetylfuran, 1-2,5-dimethyl-3-furyl ethan-1-one, 2,5-dimethyl-3-furyl methyl ketone, 1-2,5-dimethyl-3-furanyl ethanone, 1-2,5-dimethyl-3-furyl ethanone, unii-798v2t7zbv | |
CC1=CC(=C(O1)C)C(=O)C | |
138.166 | |
138.17 |
78°C (172°F) | |
50g | |
C8H10O2 | |
112094 | |
KBSVBCHYXYXDAG-UHFFFAOYSA-N | |
1-(2,5-dimethylfuran-3-yl)ethanone | |
61527 | |
98% |
Chemical Identifiers
10599-70-9 | |
138.166 | |
KBSVBCHYXYXDAG-UHFFFAOYSA-N | |
61527 | |
CC1=CC(=C(O1)C)C(=O)C |
C8H10O2 | |
MFCD00010281 | |
3-acetyl-2,5-dimethylfuran, 1-2,5-dimethylfuran-3-yl ethanone, ethanone, 1-2,5-dimethyl-3-furanyl, 3-acetyl-2,5-dimethyl furan, 2,5-dimethyl-3-acetylfuran, 1-2,5-dimethyl-3-furyl ethan-1-one, 2,5-dimethyl-3-furyl methyl ketone, 1-2,5-dimethyl-3-furanyl ethanone, 1-2,5-dimethyl-3-furyl ethanone, unii-798v2t7zbv | |
1-(2,5-dimethylfuran-3-yl)ethanone |
Safety and Handling
GHS H Statement
H301-H227-H319-H317
Toxic if swallowed.
Combustible liquid.
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger