missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetyl-2,4,6-trimethylpyridine, 98%

$78.50 - $224.73

Chemical Identifiers

CAS 56704-25-7
Molecular Formula C10H13NO
Formula Weight 163.22
MDL Number MFCD00085730
Synonym 3-Acetyl-2,4,6-collidine
Chemical Name or Material 3-Acetyl-2,4,6-trimethylpyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2713803 SDS Alfa Aesar™
H2713803
1g Each for $78.50
AAH2713806 SDS Alfa Aesar™
H2713806
5g Each for $224.73
Specifications Description & Specifications

Chemical Identifiers

56704-25-7
163.22
3-Acetyl-2,4,6-collidine
C10H13NO
MFCD00085730
3-Acetyl-2,4,6-trimethylpyridine

Specifications

56704-25-7
163.22
3-Acetyl-2,4,6-collidine
3-Acetyl-2,4,6-trimethylpyridine
C10H13NO
MFCD00085730
1g
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning