Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetyl-1-(phenylsulfonyl)pyrrole, 98%

Chemical Identifiers

Chemical Name or Material 3-Acetyl-1-(phenylsulfonyl)pyrrole
CAS 81453-98-7
Formula Weight 249.28
MDL Number MFCD00067752
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5575106

 
alfa aesar™
H5575106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5575103

 
alfa aesar™
H5575103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5575114

 
alfa aesar™
H5575114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 96° to 99°C
Formula Weight 249.28
CAS 81453-98-7
Physical Form Solid
Chemical Name or Material 3-Acetyl-1-(phenylsulfonyl)pyrrole
MDL Number MFCD00067752
Assay 98%
Quantity 5g