missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetoxybenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H54037MD

 View more versions of this product

Catalog No. AAH54037MD



Specifications

Specifications

3-Acetoxybenzonitrile
55682-11-6
MFCD00019749
#
CC(=O)OC1=CC=CC=C1C#N
161.16
161.16
250mg
C9H7NO2
2576295
XXLKCUTUGWSJJO-UHFFFAOYSA-N
(2-cyanophenyl) acetate
79791
97%

Chemical Identifiers

55682-11-6
161.16
XXLKCUTUGWSJJO-UHFFFAOYSA-N
79791
CC(=O)OC1=CC=CC=C1C#N
C9H7NO2
MFCD00019749
#
(2-cyanophenyl) acetate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents