Please login to your online account to display your discounted pricing

3-acetoxybenzofuran, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381560050

 View more versions of this product

Catalog No. AC381560050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 176.17
Physical Form Liquid
Quantity 5g
Color Clear light yellow
CAS Max % 100.0
Chemical Name or Material 3-Acetoxybenzofuran
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Molecular Formula C10H8O3
Assay Percent Range 99%
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.543 to 1.545 (20°C, 589nm)
Infrared Spectrum Authentic
MDL Number MFCD00151788
CAS 93680-80-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Acetoxybenzofuran
CAS 93680-80-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00151788
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents