Please login to your online account to display your discounted pricing

3-acetoxybenzofuran, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Acetoxybenzofuran
CAS 93680-80-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00151788
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381560050

  SDS
thermo fisher scientific
AC381560050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381560010

  SDS
thermo fisher scientific
AC381560010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 176.17
Physical Form Liquid
Quantity 5g
Color Clear light yellow
CAS Max % 100.0
Chemical Name or Material 3-Acetoxybenzofuran
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Molecular Formula C10H8O3
Assay Percent Range 99%
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.543 to 1.545 (20°C, 589nm)
Infrared Spectrum Authentic
MDL Number MFCD00151788
CAS 93680-80-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.