Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetoxybenzo[b]furan, 98%

$94.47 - $351.08

Chemical Identifiers

Chemical Name or Material 3-Acetoxybenzo[b]furan
CAS 93680-80-9
Molecular Formula C10H8O3
Formula Weight 176.17
MDL Number MFCD00151788
Beilstein 4989473
Synonym Benzo[b]furan-3-yl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1312603

  SDS
alfa aesar™
L1312603
1g Each for $94.47

AAL1312606

  SDS
alfa aesar™
L1312606
5g Each for $351.08
Description & Specifications

Specifications

Formula Weight 176.17
Chemical Name or Material 3-Acetoxybenzo[b]furan
CAS 93680-80-9
MDL Number MFCD00151788
Assay 98%
Beilstein 4989473
Quantity 1g
Molecular Formula C10H8O3
Synonym Benzo[b]furan-3-yl acetate