Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetoxybenzaldehyde, 97%

$46.64 - $163.16

Chemical Identifiers

Chemical Name or Material 3-Acetoxybenzaldehyde
CAS 34231-78-2
Molecular Formula C9H8O3
Formula Weight 164.16
MDL Number MFCD00016603
Beilstein 1941499
Synonym 3-Formylphenyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1343106

 
alfa aesar™
L1343106
5g Each for $163.16

AAL1343103

 
alfa aesar™
L1343103
1g Each for $46.64
Description & Specifications

Specifications

Formula Weight 164.16
Chemical Name or Material 3-Acetoxybenzaldehyde
Molecular Formula C9H8O3
MDL Number MFCD00016603
CAS 34231-78-2
Health Hazard 1 Warning
Beilstein 1941499
Synonym 3-Formylphenyl acetate
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 138°C (20mmHg)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 5g
Assay 97%

Applications
3-Acetoxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions. This material may darken on storage.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.