missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetoxybenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ L1343106

 View more versions of this product

Catalog No. AAL1343106



Description

Description

Applications
3-Acetoxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions. This material may darken on storage.
Specifications

Specifications

3-Acetoxybenzaldehyde
5g
C9H8O3
1941499
GVUMZPWBUAGJBP-UHFFFAOYSA-N
(3-formylphenyl) acetate
118618
97%
138°C (20mmHg)
34231-78-2
MFCD00016603
3-acetoxybenzaldehyde, benzaldehyde, 3-acetyloxy, acetoxybenzaldehyde, acetic acid 3-formylphenyl ester, m-acetoxybenzaldehyde, m-ace-toxybenzaldehyde, meta-acetoxybenzaldehyde, pubchem13227, 3-acetoxybenzaldehyde, 3-formylphenyl acetate
CC(=O)OC1=CC=CC(=C1)C=O
164.16
164.16

Chemical Identifiers

34231-78-2
164.16
GVUMZPWBUAGJBP-UHFFFAOYSA-N
118618
CC(=O)OC1=CC=CC(=C1)C=O
C9H8O3
MFCD00016603
3-acetoxybenzaldehyde, benzaldehyde, 3-acetyloxy, acetoxybenzaldehyde, acetic acid 3-formylphenyl ester, m-acetoxybenzaldehyde, m-ace-toxybenzaldehyde, meta-acetoxybenzaldehyde, pubchem13227, 3-acetoxybenzaldehyde, 3-formylphenyl acetate
(3-formylphenyl) acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents