Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetoxy-2-methylbenzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Acetoxy-2-methylbenzoyl chloride
CAS 167678-46-8
Molecular Formula C10H9ClO3
Formula Weight 212.63
MDL Number MFCD02094037
UN Number UN3261
Beilstein 8054543
Synonym 3-Acetoxy-o-toluoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1872014

  SDS
alfa aesar™
L1872014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1872006

  SDS
alfa aesar™
L1872006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.63
CAS 167678-46-8
Chemical Name or Material 3-Acetoxy-2-methylbenzoyl chloride
Molecular Formula C10H9ClO3
Assay 97%
Health Hazard 2 GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD02094037
Health Hazard 1 Danger
Synonym 3-Acetoxy-o-toluoyl chloride
Quantity 25g
Melting Point 31° to 35°C
Beilstein 8054543
UN Number UN3261
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.