missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Acetoxy-2-methylbenzoic acid, 99%

$62.46 - $218.99

Chemical Identifiers

CAS 168899-58-9
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00957176
InChI Key LHVJUPHCLWIPLZ-UHFFFAOYSA-N
Synonym 3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid, 3-acetoxy-2-methylbenzoic acid
PubChem CID 3798557
IUPAC Name 3-acetyloxy-2-methylbenzoic acid
SMILES CC1=C(C=CC=C1OC(=O)C)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1871906 View Documents Alfa Aesar™
L1871906
5g Each for $62.46
 
AAL1871914 View Documents Alfa Aesar™
L1871914
25g Each for $218.99
 
Specifications

Chemical Identifiers

168899-58-9
194.186
LHVJUPHCLWIPLZ-UHFFFAOYSA-N
3798557
CC1=C(C=CC=C1OC(=O)C)C(=O)O
C10H10O4
MFCD00957176
3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid, 3-acetoxy-2-methylbenzoic acid
3-acetyloxy-2-methylbenzoic acid

Specifications

149°C to 150°C
168899-58-9
MFCD00957176
3-acetoxy-2-methylbenzoic acid, 2-methyl-3-acetoxybenzoic acid, 3-acetoxy-2-methyl benzoic acid, 3-acetyloxy-2-methylbenzoic acid, benzoic acid, 3-acetyloxy-2-methyl, 3-acetoxy-2-methyl-benzoic acid, acmc-209dz2, ksc495o2n, 3-acetyloxy-2-methyl-benzoic acid, 3-acetoxy-2-methylbenzoic acid
CC1=C(C=CC=C1OC(=O)C)C(=O)O
194.186
194.19
3-Acetoxy-2-methylbenzoic acid
5g
C10H10O4
3271605
LHVJUPHCLWIPLZ-UHFFFAOYSA-N
3-acetyloxy-2-methylbenzoic acid
3798557
99%
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning