Learn More
3-Acetamidophenol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 102320250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Acetamidophenol | |
621-42-1 | |
100.0 | |
Gray or Tan to White | |
99% min. (GC) | |
C8H9NO2 | |
MFCD00002263 | |
13, 415 | |
Solubility in water: freely soluble in hot water | |
CC(=O)NC1=CC(=CC=C1)O | |
151.16 | |
CHEBI:76987 | |
99+% |
99+% | |
99.0 | |
146°C to 149°C | |
Authentic | |
Glass bottle | |
CH3CONHC6H4OH | |
25 g | |
3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid | |
QLNWXBAGRTUKKI-UHFFFAOYSA-N | |
N-(3-hydroxyphenyl)acetamide | |
12124 | |
151.16 | |
Crystalline Powder, Crystals or Needles |
Chemical Identifiers
621-42-1 | |
151.16 | |
QLNWXBAGRTUKKI-UHFFFAOYSA-N | |
12124 | |
N-(3-hydroxyphenyl)acetamide |
C8H9NO2 | |
MFCD00002263 | |
3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid | |
CHEBI:76987 | |
CC(=O)NC1=CC(=CC=C1)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 210-687-0
RTECSNumber : AE4100000
TSCA : TSCA
RUO – Research Use Only