Please login to your online account to display your discounted pricing

3-Acetamidophenol 99+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  102321000

 View more versions of this product

Catalog No. AC102321000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 151.16
Physical Form Crystals, Crystalline Powder or Needles
Quantity 100g
Color Off-white to tan or light gray
RTECS Number AE4100000
EINECS Number 210-687-0
TSCA TSCA
Melting Point 146° to 149°C
Beilstein 13, 415
CAS Max % 100.0
CAS Min % 99.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 LIGHT SENSITIVE
Assay Percent Range 99+%
Linear Formula CH3CONHC6H4OH
CAS 621-42-1
Molecular Formula C8H9NO2
Chemical Name or Material 3-Acetamidophenol
Name Note 99+%
MDL Number MFCD00002263
Synonym m-Hydroxyacetanilide
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Acetamidophenol
Name Note 99+%
CAS 621-42-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C8H9NO2
Linear Formula CH3CONHC6H4OH
Formula Weight 151.16
MDL Number MFCD00002263
Synonym m-Hydroxyacetanilide
Accessories

LIGHT SENSITIVE

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents