missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Acetamidophenol, 99+%, ACROS Organics™
$41.39 - $41.39
Chemical Identifiers
CAS | 621-42-1 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.16 |
MDL Number | MFCD00002263 |
InChI Key | QLNWXBAGRTUKKI-UHFFFAOYSA-N |
Synonym | 3'-hydroxyacetanilide, 3-acetamidophenol, 3-acetylamino phenol, 3-hydroxyacetanilide, m-acetamidophenol, m-hydroxyacetanilide, metacetamol, metalid, n-3-hydroxyphenyl acetamide, n-acetyl-m-aminophenol |
PubChem CID | 12124 |
ChEBI | CHEBI:76987 |
IUPAC Name | N-(3-hydroxyphenyl)acetamide |
SMILES | CC(=O)NC1=CC(=CC=C1)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC102320250
|
Thermo Fisher Scientific
102320250 |
25g | Glass bottle |
Each for $41.39
|
|
AC102321000
|
Thermo Fisher Scientific
102321000 |
100g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
621-42-1 | |
151.16 | |
QLNWXBAGRTUKKI-UHFFFAOYSA-N | |
12124 | |
N-(3-hydroxyphenyl)acetamide |
C8H9NO2 | |
MFCD00002263 | |
3'-hydroxyacetanilide, 3-acetamidophenol, 3-acetylamino phenol, 3-hydroxyacetanilide, m-acetamidophenol, m-hydroxyacetanilide, metacetamol, metalid, n-3-hydroxyphenyl acetamide, n-acetyl-m-aminophenol | |
CHEBI:76987 | |
CC(=O)NC1=CC(=CC=C1)O |
Specifications
Authentic | |
Gray or Tan to White | |
25g | |
99.0 | |
99% min. (GC) | |
CH3CONHC6H4OH | |
13, 415 | |
Solubility in water: freely soluble in hot water | |
CC(=O)NC1=CC(=CC=C1)O | |
151.16 | |
CHEBI:76987 | |
Crystals, Crystalline Powder or Needles | |
3-Acetamidophenol, 99+% |
Glass bottle | |
146°C to 149°C | |
621-42-1 | |
100.0 | |
C8H9NO2 | |
MFCD00002263 | |
3'-hydroxyacetanilide, 3-acetamidophenol, 3-acetylamino phenol, 3-hydroxyacetanilide, m-acetamidophenol, m-hydroxyacetanilide, metacetamol, metalid, n-3-hydroxyphenyl acetamide, n-acetyl-m-aminophenol | |
QLNWXBAGRTUKKI-UHFFFAOYSA-N | |
N-(3-hydroxyphenyl)acetamide | |
12124 | |
151.16 | |
99+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 210-687-0
RTECSNumber : AE4100000
TSCA : TSCA