Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Acetamidomethyl)pyridine, 97%

$182.32 - $182.32

Chemical Identifiers

Chemical Name or Material 3-(Acetamidomethyl)pyridine
CAS 22977-34-0
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00495392
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2186209

  SDS
alfa aesar™
B2186209
10g Each for $182.32
Description & Specifications

Specifications

Formula Weight 150.18
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
CAS 22977-34-0
Boiling Point 152° to 154°C (0.8mmHg)
Quantity 10g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H10N2O
Health Hazard 1 Warning
Chemical Name or Material 3-(Acetamidomethyl)pyridine
MDL Number MFCD00495392
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.