missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Acetamidomethyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H53252MD

 View more versions of this product

Catalog No. AAH53252MD



Description

Description

Applications
3-(Acetamidomethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-(Acetamidomethyl)benzeneboronic acid
250mg
C9H12BNO3
3-acetamidomethyl phenyl boronic acid, 3-acetamidomethylphenylboronic acid, 3-acetamidomethyl phenyl boronic acid, 3-acetylaminomethyl benzeneboronic acid, 3-acetamidomethyl phenylboronic acid, 3-acetamidomethyl benzeneboronic acid, 3-acetamidomethyl benzeneboronic acid, boronic acid,b-3-acetylamino methyl phenyl, pubchem14581, acmc-209q1y
B(C1=CC(=CC=C1)CNC(=O)C)(O)O
193.009
193.01
253°C to 255°C
850568-42-2
MFCD06659819
IXRGNMIMGAJHEG-UHFFFAOYSA-N
[3-(acetamidomethyl)phenyl]boronic acid
22309455
97%

Chemical Identifiers

850568-42-2
193.009
IXRGNMIMGAJHEG-UHFFFAOYSA-N
22309455
B(C1=CC(=CC=C1)CNC(=O)C)(O)O
C9H12BNO3
MFCD06659819
3-acetamidomethyl phenyl boronic acid, 3-acetamidomethylphenylboronic acid, 3-acetamidomethyl phenyl boronic acid, 3-acetylaminomethyl benzeneboronic acid, 3-acetamidomethyl phenylboronic acid, 3-acetamidomethyl benzeneboronic acid, 3-acetamidomethyl benzeneboronic acid, boronic acid,b-3-acetylamino methyl phenyl, pubchem14581, acmc-209q1y
[3-(acetamidomethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents