missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Acetamidoacetophenone, 98%

Manufacturer:  Alfa Aesar™ A1218814

 View more versions of this product

Catalog No. AAA1218814


 

Specifications

Specifications

3'-Acetamidoacetophenone
98%
MFCD00032278
3'-Acetylacetanilide; N,3-Diacetylaniline
CC(=O)C1=CC(=CC=C1)NC(=O)C
177.203
177.2
25g
7463-31-2
C10H11NO2
2363443
AFZTYHRVDOKRKV-UHFFFAOYSA-N
N-(3-acetylphenyl)acetamide
346202
125°C to 129°C

Chemical Identifiers

7463-31-2
177.203
AFZTYHRVDOKRKV-UHFFFAOYSA-N
346202
C10H11NO2
MFCD00032278
3'-Acetylacetanilide; N,3-Diacetylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents