Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,9-Dodecadiyne, 97%

$72.49 - $249.36

Chemical Identifiers

Chemical Name or Material 3,9-Dodecadiyne
CAS 61827-89-2
Molecular Formula C12H18
Formula Weight 162.28
MDL Number MFCD00041637
Beilstein 1698817
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0986306

 
alfa aesar™
L0986306
5g Each for $72.49

AAL0986314

 
alfa aesar™
L0986314
25g Each for $249.36
Description & Specifications

Specifications

Formula Weight 162.28
Chemical Name or Material 3,9-Dodecadiyne
Assay 97%
Molecular Formula C12H18
MDL Number MFCD00041637
CAS 61827-89-2
Beilstein 1698817
Boiling Point 55° to 58°C (0.5mmHg)
Quantity 5g