missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,9-Dodecadiyne, 97%

Manufacturer:  Alfa Aesar™ L0986306

 View more versions of this product

Catalog No. AAL0986306



Specifications

Specifications

3,9-Dodecadiyne
5g
C12H18
1698817
SLQXYOMUNKJHRV-UHFFFAOYSA-N
dodeca-3,9-diyne
143841
97%
55°C to 58°C (0.5mmHg)
61827-89-2
MFCD00041637
3,9-dodecadiyne, acmc-209mwh, 3,9-dodecadiyne, 3,9-dodecadiyne,
CCC#CCCCCC#CCC
162.276
162.28

Chemical Identifiers

61827-89-2
162.276
SLQXYOMUNKJHRV-UHFFFAOYSA-N
143841
CCC#CCCCCC#CCC
C12H18
MFCD00041637
3,9-dodecadiyne, acmc-209mwh, 3,9-dodecadiyne, 3,9-dodecadiyne,
dodeca-3,9-diyne
Documents