missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-3,9-Dibromocamphor 97.0+%, TCI America™
Supplier: TCI America D271525G
Specifications
| (+)-3,9-Dibromocamphor | |
| White | |
| MFCD00167983 | |
| DCDNKSJBRIJYEC-MZIHZIJQSA-N | |
| (1S,2S,4R)-2-bromo-7-(bromomethyl)-4,7-dimethylbicyclo[2.2.1]heptan-3-one | |
| 13783905 | |
| ≥97.0% (GC) |
| 10293-10-4 | |
| C10H14Br2O | |
| 25 g | |
| CC12CCC(C1(C)CBr)C(C2=O)Br | |
| 310.029 | |
| 310.03 | |
| Crystalline Powder |
Chemical Identifiers
| 10293-10-4 | |
| 310.029 | |
| DCDNKSJBRIJYEC-MZIHZIJQSA-N | |
| (1S,2S,4R)-2-bromo-7-(bromomethyl)-4,7-dimethylbicyclo[2.2.1]heptan-3-one |
| C10H14Br2O | |
| MFCD00167983 | |
| 13783905 | |
| CC12CCC(C1(C)CBr)C(C2=O)Br |
Safety and Handling
TSCA : No