missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,8-Dimethoxy-2-naphthaldehyde, 99%

$334.64 - $334.64

Chemical Identifiers

CAS 374538-05-3
Molecular Formula C13H12O3
Molecular Weight (g/mol) 216.236
MDL Number MFCD02093990
InChI Key YBCQBSORAFCHGW-UHFFFAOYSA-N
Synonym 3,8-dimethoxy-2-naphthaldehyde, acmc-20anbf
PubChem CID 7010328
IUPAC Name 3,8-dimethoxynaphthalene-2-carbaldehyde
SMILES COC1=CC=CC2=CC(=C(C=C21)C=O)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1813003 View Documents Alfa Aesar™
L1813003
1g Each for $334.64
 
Specifications

Chemical Identifiers

374538-05-3
216.236
YBCQBSORAFCHGW-UHFFFAOYSA-N
7010328
COC1=CC=CC2=CC(=C(C=C21)C=O)OC
C13H12O3
MFCD02093990
3,8-dimethoxy-2-naphthaldehyde, acmc-20anbf
3,8-dimethoxynaphthalene-2-carbaldehyde

Specifications

136°C to 138°C
374538-05-3
MFCD02093990
YBCQBSORAFCHGW-UHFFFAOYSA-N
3,8-dimethoxynaphthalene-2-carbaldehyde
7010328
99%
1g
C13H12O3
3,8-dimethoxy-2-naphthaldehyde, acmc-20anbf
COC1=CC=CC2=CC(=C(C=C21)C=O)OC
216.236
216.24
3,8-Dimethoxy-2-naphthaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning