Learn More
3,8-Diamino-6-phenylphenanthridine, 98%, Thermo Scientific Chemicals
$135.44 - $504.42
Chemical Identifiers
CAS | 52009-64-0 |
---|---|
Molecular Formula | C19H15N3 |
Molecular Weight (g/mol) | 285.35 |
MDL Number | MFCD00075152 |
InChI Key | CPNAVTYCORRLMH-UHFFFAOYSA-N |
Synonym | 3,8-diamino-6-phenylphenanthridine, 6-phenylphenathridine-3,8-diamine, 3,8-dapp, 3,8-diamino-6-phenyl-phenanthridine, acmc-1aycj, 6-phenyl-3,8-phenanthridinediamine, 3,8-diamino-6-phenyl phenanthridine, 3,8-phenanthridinediamine,6-phenyl, 3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine, |
PubChem CID | 104052 |
IUPAC Name | 6-phenylphenanthridine-3,8-diamine |
SMILES | C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2404903
|
Thermo Scientific Chemicals
B2404903 |
1 g |
Each for $135.44
|
|
|||||
AAB2404906
|
Thermo Scientific Chemicals
B2404906 |
5 g |
Each for $504.42
|
|
|||||
Description
3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,8-Diamino-6-phenylphenanthridine is used in the synthesis of rigid polyamides. It has anti-tumor and anti-viral properties.
Solubility
Soluble in chloroform, dichloromethane and methanol.
Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Chemical Identifiers
52009-64-0 | |
285.35 | |
CPNAVTYCORRLMH-UHFFFAOYSA-N | |
104052 | |
C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N |
C19H15N3 | |
MFCD00075152 | |
3,8-diamino-6-phenylphenanthridine, 6-phenylphenathridine-3,8-diamine, 3,8-dapp, 3,8-diamino-6-phenyl-phenanthridine, acmc-1aycj, 6-phenyl-3,8-phenanthridinediamine, 3,8-diamino-6-phenyl phenanthridine, 3,8-phenanthridinediamine,6-phenyl, 3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine, | |
6-phenylphenanthridine-3,8-diamine |
Specifications
52009-64-0 | |
C19H15N3 | |
1 g | |
Soluble in chloroform,dichloromethane and methanol. | |
C1=CC=C(C=C1)C2=C3C=C(C=CC3=C4C=CC(=CC4=N2)N)N | |
285.35 | |
285.35 | |
3,8-Diamino-6-phenylphenanthridine |
196°C to 198°C | |
MFCD00075152 | |
3,8-diamino-6-phenylphenanthridine, 6-phenylphenathridine-3,8-diamine, 3,8-dapp, 3,8-diamino-6-phenyl-phenanthridine, acmc-1aycj, 6-phenyl-3,8-phenanthridinediamine, 3,8-diamino-6-phenyl phenanthridine, 3,8-phenanthridinediamine,6-phenyl, 3,8-diamino-6-phenylphenanthridine6-phenyl-3,8-phenanthridinediamine, | |
CPNAVTYCORRLMH-UHFFFAOYSA-N | |
6-phenylphenanthridine-3,8-diamine | |
104052 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 257-602-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only