missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,7-Dimethyl-1-octanol 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC392620010

 View more versions of this product

Catalog No. AC392620010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
158.28
Clear colorless
100.0
Glass bottle
(CH3)2CH(CH2)3CH(CH3)(CH2)2OH
C10H22O
3, 7-Dimethyl-1-octanol
Authentic
Liquid
1L
0.8280g/mL
94.0
1.4340 to 1.4360 (20°C, 589nm)
95%
MFCD00002936
106-21-8

Chemical Identifiers

106-21-8
100.0
C10H22O
158.28
3, 7-Dimethyl-1-octanol
94.0
95%
(CH3)2CH(CH2)3CH(CH3)(CH2)2OH
MFCD00002936
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning