missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(6-Oxo-1,6-dihydro-3-pyridazinyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5466206

 View more versions of this product

Catalog No. AAH5466206



Description

Description

Specifications

Specifications

3-(6-Oxo-1,6-dihydro-3-pyridazinyl)benzonitrile
52240-08-1
MFCD16482247
INFIYPRKBSENAH-UHFFFAOYSA-N
3-(6-oxo-1H-pyridazin-3-yl)benzonitrile
20326520
97%
5g
C11H7N3O
3-6-oxo-1,6-dihydro-3-pyridazinyl benzonitrile, 3-6-oxo-1,6-dihydro-pyridazin-3-yl-benzonitrile, 3-6-oxo-1,6-dihydropyridazin-3-yl benzonitrile, 3-6-oxo-1,6-dihydropyridazin-3-yl-benzonitrile, 3-6-oxo-1h-pyridazin-3-yl benzonitrile, benzonitrile, 3-1,6-dihydro-6-oxo-3-pyridazinyl
C1=CC(=CC(=C1)C2=NNC(=O)C=C2)C#N
197.197
197.19

Chemical Identifiers

52240-08-1
197.197
INFIYPRKBSENAH-UHFFFAOYSA-N
20326520
C1=CC(=CC(=C1)C2=NNC(=O)C=C2)C#N
C11H7N3O
MFCD16482247
3-6-oxo-1,6-dihydro-3-pyridazinyl benzonitrile, 3-6-oxo-1,6-dihydro-pyridazin-3-yl-benzonitrile, 3-6-oxo-1,6-dihydropyridazin-3-yl benzonitrile, 3-6-oxo-1,6-dihydropyridazin-3-yl-benzonitrile, 3-6-oxo-1h-pyridazin-3-yl benzonitrile, benzonitrile, 3-1,6-dihydro-6-oxo-3-pyridazinyl
3-(6-oxo-1H-pyridazin-3-yl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents