Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dimethyl-3-heptanol, 99%

$100.51 - $1,671.66

Chemical Identifiers

Chemical Name or Material 3,6-Dimethyl-3-heptanol
CAS 1573-28-0
Molecular Formula C9H20O
Formula Weight 144.25
MDL Number MFCD00021828
Synonym Ethyl isopentyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2028509

  SDS
alfa aesar™
B2028509
10g Each for $100.51

AAB2028530

  SDS
alfa aesar™
B2028530
250g Each for $1,671.66

AAB2028518

  SDS
alfa aesar™
B2028518
50g Each for $403.30
Description & Specifications

Specifications

Formula Weight 144.25
Chemical Name or Material 3,6-Dimethyl-3-heptanol
Assay 99%
Molecular Formula C9H20O
MDL Number MFCD00021828
CAS 1573-28-0
Synonym Ethyl isopentyl methyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.