Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dimethyl-3,6-octanediol, (+/-) + meso, 97%

$54.91 - $54.91

Chemical Identifiers

Chemical Name or Material 3,6-Dimethyl-3,6-octanediol
CAS 78-65-9
Name Note (+/-) + meso
Molecular Formula C10H22O2
Formula Weight 174.28g/mol
MDL Number MFCD00021827
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2073803

 
alfa aesar™
B20738-03
1g Each for $54.91
Description & Specifications

Specifications

Melting Point 45° to 48°C
Quantity 1g
Odor Odorless
Chemical Name or Material 3,6-Dimethyl-3,6-octanediol
Name Note (+/-) + meso
CAS 78-65-9
Assay 97%
Molecular Formula C10H22O2
Formula Weight 174.28g/mol
MDL Number MFCD00021827