Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dimethoxypyridazine-4-carboxylic acid, 96%

$150.68 - $456.55

Chemical Identifiers

Chemical Name or Material 3,6-Dimethoxypyridazine-4-carboxylic acid
CAS 89694-24-6
Molecular Formula C7H8N2O4
Formula Weight 184.15
MDL Number MFCD03426073
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2687703

  SDS
alfa aesar™
H2687703
1g Each for $150.68

AAH2687706

  SDS
alfa aesar™
H2687706
5g Each for $456.55
Description & Specifications

Specifications

Formula Weight 184.15
Health Hazard 1 Warning
Molecular Formula C7H8N2O4
Assay 96%
CAS 89694-24-6
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3,6-Dimethoxypyridazine-4-carboxylic acid
MDL Number MFCD03426073
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.