Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dichloropyridazine-4-carboxylic acid, 98%

$28.01 - $240.12

Chemical Identifiers

Chemical Name or Material 3,6-Dichloropyridazine-4-carboxylic acid
CAS 51149-08-7
Molecular Formula C5H2Cl2N2O2
Formula Weight 192.99
MDL Number MFCD00011575
Beilstein 610056
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1041806

  SDS
alfa aesar™
L1041806
5g Each for $240.12

AAL10418MD

  SDS
alfa aesar™
L10418MD
250mg Each for $28.01
Description & Specifications

Specifications

Formula Weight 192.99
Chemical Name or Material 3,6-Dichloropyridazine-4-carboxylic acid
Assay 98%
Molecular Formula C5H2Cl2N2O2
MDL Number MFCD00011575
CAS 51149-08-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 610056
Melting Point 137° to 140°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.