missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,6-Dichloro-2-(trifluoromethyl)pyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451380010
Specifications
3, 6-Dichloro-2-(trifluoromethyl)pyridine | |
88°C (16.0mmHg) | |
89719-91-5 | |
C6H2Cl2F3N | |
WDFIJOQQKFWJEA-UHFFFAOYSA-N | |
3,6-dichloro-2-(trifluoromethyl)pyridine | |
20213813 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
2,5-dichloro-6-trifluoromethyl pyridine, 3,6-dichloro-2-trifluoromethyl pyridine, acmc-20a3gu, pyridine, 3,6-dichloro-2-trifluoromethyl | |
C1=CC(=NC(=C1Cl)C(F)(F)F)Cl | |
215.99 | |
215.99 |
Chemical Identifiers
89719-91-5 | |
215.99 | |
2,5-dichloro-6-trifluoromethyl pyridine, 3,6-dichloro-2-trifluoromethyl pyridine, acmc-20a3gu, pyridine, 3,6-dichloro-2-trifluoromethyl | |
3,6-dichloro-2-(trifluoromethyl)pyridine |
C6H2Cl2F3N | |
WDFIJOQQKFWJEA-UHFFFAOYSA-N | |
20213813 | |
C1=CC(=NC(=C1Cl)C(F)(F)F)Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning