missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,6-Dichloro-2-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC451380050

 View more versions of this product

Catalog No. AC451380050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 6-Dichloro-2-(trifluoromethyl)pyridine
5g
96% min. (GC)
2,5-dichloro-6-trifluoromethyl pyridine, 3,6-dichloro-2-trifluoromethyl pyridine, acmc-20a3gu, pyridine, 3,6-dichloro-2-trifluoromethyl
C1=CC(=NC(=C1Cl)C(F)(F)F)Cl
215.99
215.99
88°C (16.0mmHg)
89719-91-5
C6H2Cl2F3N
WDFIJOQQKFWJEA-UHFFFAOYSA-N
3,6-dichloro-2-(trifluoromethyl)pyridine
20213813
97%

Chemical Identifiers

89719-91-5
215.99
2,5-dichloro-6-trifluoromethyl pyridine, 3,6-dichloro-2-trifluoromethyl pyridine, acmc-20a3gu, pyridine, 3,6-dichloro-2-trifluoromethyl
3,6-dichloro-2-(trifluoromethyl)pyridine
C6H2Cl2F3N
WDFIJOQQKFWJEA-UHFFFAOYSA-N
20213813
C1=CC(=NC(=C1Cl)C(F)(F)F)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

SDS Documents