Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dichloro-2-fluorobenzylamine, 97%

Chemical Identifiers

Chemical Name or Material 3,6-Dichloro-2-fluorobenzylamine
Molecular Formula C7H6Cl2FN
Formula Weight 194.04
MDL Number MFCD09025386
UN Number UN3259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3213203

 
alfa aesar™
H3213203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32132MD

 
alfa aesar™
H32132MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.04
Chemical Name or Material 3,6-Dichloro-2-fluorobenzylamine
Assay 97%
Molecular Formula C7H6Cl2FN
MDL Number MFCD09025386
UN Number UN3259
Melting Point 44° to 47°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.