Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dichloro-2-fluorobenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 3,6-Dichloro-2-fluorobenzonitrile
Molecular Formula C7H2Cl2FN
Formula Weight 190
MDL Number MFCD09025385
UN Number UN3439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3275203

 
alfa aesar™
H3275203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3275206

 
alfa aesar™
H3275206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190
Chemical Name or Material 3,6-Dichloro-2-fluorobenzonitrile
Assay 97%
Molecular Formula C7H2Cl2FN
MDL Number MFCD09025385
UN Number UN3439
Melting Point 61° to 64°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 1g

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.