Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dichloro-2-fluoroaniline, 97%

Chemical Identifiers

Chemical Name or Material 3,6-Dichloro-2-fluoroaniline
CAS 348-64-1
Molecular Formula C6H4Cl2FN
Formula Weight 180.01
MDL Number MFCD09832331
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3233303

 
alfa aesar™
H3233303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3233306

 
alfa aesar™
H3233306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.01
Chemical Name or Material 3,6-Dichloro-2-fluoroaniline
Assay 97%
Molecular Formula C6H4Cl2FN
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
MDL Number MFCD09832331
CAS 348-64-1
UN Number UN2811
Melting Point 54° to 56°C
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.