missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dibromopyridazine, 95%

Manufacturer:  Alfa Aesar™ H6122306

 View more versions of this product

Catalog No. AAH6122306



Specifications

Specifications

3,6-Dibromopyridazine
117°C to 121°C
17973-86-3
MFCD00233947
VQAFMTSSCUETHA-UHFFFAOYSA-N
3,6-dibromopyridazine
248852
Solid
White to Yellow
5g
C4H2Br2N2
3,6-dibromo pyridazine, 3,6-dibromo-1,2-diazine, 3,6-dibromo-pyridazine, 3,6-dibromopyridazide, 3,6-dibromopyridazine, pubchem12385, pyridazine, 3,6-dibromo, pyridazine,3,6-dibromo, synquest 3h32-9-t3
C1=CC(=NN=C1Br)Br
237.882
237.88
95%

Chemical Identifiers

17973-86-3
237.882
VQAFMTSSCUETHA-UHFFFAOYSA-N
248852
C1=CC(=NN=C1Br)Br
C4H2Br2N2
MFCD00233947
3,6-dibromo pyridazine, 3,6-dibromo-1,2-diazine, 3,6-dibromo-pyridazine, 3,6-dibromopyridazide, 3,6-dibromopyridazine, pubchem12385, pyridazine, 3,6-dibromo, pyridazine,3,6-dibromo, synquest 3h32-9-t3
3,6-dibromopyridazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents