missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dibromopyrazine-2-carboxamide, 97%

$127.36 - $1,688.62

Chemical Identifiers

CAS 1301613-77-3
Molecular Formula C5H3Br2N3O
Molecular Weight (g/mol) 280.907
InChI Key TWCACYWIPUIXAR-UHFFFAOYSA-N
PubChem CID 58269823
IUPAC Name 3,6-dibromopyrazine-2-carboxamide
SMILES C1=C(N=C(C(=N1)Br)C(=O)N)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63856MD View Documents Alfa Aesar™
H63856MD
250mg Each for $127.36
 
AAH6385603 View Documents Alfa Aesar™
H6385603
1g Each for $405.58
 
AAH6385606 View Documents Alfa Aesar™
H6385606
5g Each for $1,688.62
 
Specifications

Chemical Identifiers

1301613-77-3
280.907
58269823
C1=C(N=C(C(=N1)Br)C(=O)N)Br
C5H3Br2N3O
TWCACYWIPUIXAR-UHFFFAOYSA-N
3,6-dibromopyrazine-2-carboxamide

Specifications

250mg
C5H3Br2N3O
C1=C(N=C(C(=N1)Br)C(=O)N)Br
280.907
280.9
97%
1301613-77-3
TWCACYWIPUIXAR-UHFFFAOYSA-N
3,6-dibromopyrazine-2-carboxamide
58269823
Solid
3,6-Dibromopyrazine-2-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning