missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dibromo-9-phenylcarbazole, 98%

$22.36 - $70.31

Chemical Identifiers

CAS 57103-20-5
Molecular Formula C18H11Br2N
Molecular Weight (g/mol) 401.101
MDL Number MFCD08276338
InChI Key JBWRZTKHMKVFMQ-UHFFFAOYSA-N
Synonym 3,6-dibromo-9-phenyl-9h-carbazole, 9h-carbazole, 3,6-dibromo-9-phenyl, 3,6-dibromo-9-phenyl-carbazole, ksc268e0n, 3,6-dibromo-9-phenylarbazole, n-phenyl-3,6-dibromocarbazole, acmc-1b118, 3,6-dibromo-9-phenylcarbazole
PubChem CID 11025791
IUPAC Name 3,6-dibromo-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60029MD View Documents Alfa Aesar™
H60029MD
250mg Each for $22.36
 
AAH6002903 View Documents Alfa Aesar™
H6002903
1g Each for $70.31
 
Specifications

Chemical Identifiers

57103-20-5
401.101
JBWRZTKHMKVFMQ-UHFFFAOYSA-N
11025791
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br
C18H11Br2N
MFCD08276338
3,6-dibromo-9-phenyl-9h-carbazole, 9h-carbazole, 3,6-dibromo-9-phenyl, 3,6-dibromo-9-phenyl-carbazole, ksc268e0n, 3,6-dibromo-9-phenylarbazole, n-phenyl-3,6-dibromocarbazole, acmc-1b118, 3,6-dibromo-9-phenylcarbazole
3,6-dibromo-9-phenylcarbazole

Specifications

160°C to 164°C
57103-20-5
MFCD08276338
JBWRZTKHMKVFMQ-UHFFFAOYSA-N
3,6-dibromo-9-phenylcarbazole
11025791
98%
250mg
C18H11Br2N
3,6-dibromo-9-phenyl-9h-carbazole, 9h-carbazole, 3,6-dibromo-9-phenyl, 3,6-dibromo-9-phenyl-carbazole, ksc268e0n, 3,6-dibromo-9-phenylarbazole, n-phenyl-3,6-dibromocarbazole, acmc-1b118, 3,6-dibromo-9-phenylcarbazole
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br
401.101
401.09
3,6-Dibromo-9-phenylcarbazole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement
P280-P301+P310-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger