Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,6-Dibromo-2-methylpyridine, 95%

$50.62 - $116.03

Chemical Identifiers

Chemical Name or Material 3,6-Dibromo-2-methylpyridine
CAS 39919-65-8
Molecular Formula C6H5Br2N
Formula Weight 250.92
MDL Number MFCD06254589
Synonym 3,6-Dibromo-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2739903

  SDS
alfa aesar™
H2739903
1g Each for $116.03

AAH27399MD

  SDS
alfa aesar™
H27399MD
250mg Each for $50.62
Description & Specifications

Specifications

Formula Weight 250.92
Chemical Name or Material 3,6-Dibromo-2-methylpyridine
Melting Point 36° to 39°C
Assay 95%
Quantity 1g
Molecular Formula C6H5Br2N
MDL Number MFCD06254589
CAS 39919-65-8
Synonym 3,6-Dibromo-2-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.