Please login to your online account to display your discounted pricing

3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane, 41% solution in aromatic free mi, ACROS Organics™

$94.87 - $94.87

Chemical Identifiers

Chemical Name or Material 3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane
Name Note 41% Solution in Aromatic Free Min. Spirit
CAS 24748-23-0
CAS Min % 40.0
CAS Max % 42.0
CAS 78-93-3
CAS Min % 58.0
CAS Max % 60.0
Assay 7.3 to 7.6% (total active oxygen)
Molecular Formula C12H24O6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC349941000

  SDS
thermo fisher scientific
349941000
Plastic bottle 100g Each for $94.87

AC349940050

  SDS
thermo fisher scientific
AC349940050
Plastic bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 264.32
Quantity 100g
Color Clear
TSCA TSCA
Assay 7.3 to 7.6% (total active oxygen)
Density 0.8750g/mL
CAS Min % 40.0
CAS Max % 42.0
CAS Min % 58.0
CAS Max % 60.0
Chemical Name or Material 3,6,9-Triethyl-3,6,9-trimethyl-1,4,7-triperoxonane
Name Note 41% Solution in Aromatic Free Min. Spirit
Molecular Formula C12H24O6
MDL Number MFCD02685709
CAS 24748-23-0
Synonym Trigonox(r) 30
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Heating may cause a fire or explosion.
Flammable liquid and vapor.
CAS 78-93-3
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle

Radical initiators

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Causes skin irritation.
May be fatal if swallowed and enters airways.
Heating may cause a fire or explosion.
Flammable liquid and vapor.