Please login to your online account to display your discounted pricing

3(5)-Phenyl-1H-pyrazole, 99+%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC296030100

 View more versions of this product

Catalog No. AC296030100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder
Formula Weight 144.18
Quantity 10g
Color White to yellow-orange
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Melting Point 74° to 80°C
Color Index No. 3(5)-Phenyl-1H-pyrazole
Beilstein 23, IV, 1238
CAS Max % 100.0
CAS Min % 99.0
Infrared Spectrum Authentic
Health Hazard 1 Warning
Chemical Name or Material 3(5)-Phenyl-1H-pyrazole
Assay Percent Range 99+%
Packaging Glass bottle
CAS 2458-26-6
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Molecular Formula C9H8N2

Specifications

Chemical Name or Material 3(5)-Phenyl-1H-pyrazole
CAS 2458-26-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H8N2
Formula Weight 144.18
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.

GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents