Please login to your online account to display your discounted pricing

3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride, Tech., Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC39103DA

 View more versions of this product

Catalog No. CC39103DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 258.68
Quantity 1g
Assay Tech.
CAS Min % 80.0
CAS Max % 100.0
Molecular Formula C9H7ClN2O3S
MDL Number MFCD06797486
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzenesulfonyl chloride
Packaging Amber Glass Bottle
CAS 10185-62-3

Specifications

Chemical Name or Material 3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzenesulfonyl chloride
CAS 10185-62-3
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay Tech.
Molecular Formula C9H7ClN2O3S
Formula Weight 258.68
MDL Number MFCD06797486
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.