missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Lutidine, 99%

Manufacturer:  Alfa Aesar™ A14877AP

 View more versions of this product

Catalog No. AAA14877AP



Specifications

Specifications

3,5-Lutidine
Unpleasant
-9°C
591-22-0
MFCD00006404
105682
Soluble in water
CC1=CC(=CN=C1)C
107.156
107.16
53°C (127°F)
169°C to 170°C
500mL
C7H9N
UN1992
3,5-lutidine, pyridine, 3,5-dimethyl, unii-6q4ypz045v, 3,5-dimethyl pyridine, 3,5 lutidine, 3,5-lutdcine, 3,5-dimethylpyridin, 3,5-dimethyl-pyridine, acmc-1axev, dsstox_cid_30825
HWWYDZCSSYKIAD-UHFFFAOYSA-N
3,5-dimethylpyridine
11565
99%

Chemical Identifiers

591-22-0
107.156
HWWYDZCSSYKIAD-UHFFFAOYSA-N
11565
CC1=CC(=CN=C1)C
C7H9N
MFCD00006404
3,5-lutidine, pyridine, 3,5-dimethyl, unii-6q4ypz045v, 3,5-dimethyl pyridine, 3,5 lutidine, 3,5-lutdcine, 3,5-dimethylpyridin, 3,5-dimethyl-pyridine, acmc-1axev, dsstox_cid_30825
3,5-dimethylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H226-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P304+P340-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA

SDS Documents