Learn More
3,5-Heptanedione, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 7424-54-6 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00015186 |
InChI Key | DGCTVLNZTFDPDJ-UHFFFAOYSA-N |
Synonym | 3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc |
PubChem CID | 81923 |
IUPAC Name | heptane-3,5-dione |
SMILES | CCC(=O)CC(=O)CC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC411260050
|
Thermo Scientific Chemicals
411260050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC411260250
|
Thermo Scientific Chemicals
411260250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7424-54-6 | |
128.17 | |
DGCTVLNZTFDPDJ-UHFFFAOYSA-N | |
81923 | |
CCC(=O)CC(=O)CC |
C7H12O2 | |
MFCD00015186 | |
3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc | |
heptane-3,5-dione |
Specifications
7424-54-6 | |
100.0 | |
172°C to 173°C (711.0mmHg) | |
Authentic | |
Glass bottle | |
1.4540 to 1.4570 (20°C, 589nm) | |
MFCD00015186 | |
0.945 | |
DGCTVLNZTFDPDJ-UHFFFAOYSA-N | |
heptane-3,5-dione | |
81923 | |
99% |
98.5 | |
0.9450g/mL | |
57°C | |
98.5% min. (GC) | |
C7H12O2 | |
CH3CH2COCH2COCH2CH3 | |
5g | |
3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc | |
CCC(=O)CC(=O)CC | |
128.17 | |
128.17 | |
3, 5-Heptanedione |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 231-054-5
TSCA : TSCA
RUO â Research Use Only