Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Diphenyl-1H-pyrazole, 98+%

$95.54 - $390.26

Chemical Identifiers

Chemical Name or Material 3,5-Diphenyl-1H-pyrazole
CAS 1145-01-3
Molecular Formula C15H12N2
Formula Weight 220.28
MDL Number MFCD00039675
Beilstein 158264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0490406

  SDS
alfa aesar™
L0490406
5g Each for $95.54

AAL0490414

  SDS
alfa aesar™
L0490414
25g Each for $390.26
Description & Specifications

Specifications

Formula Weight 220.28
Chemical Name or Material 3,5-Diphenyl-1H-pyrazole
MDL Number MFCD00039675
Assay ≥98%
Molecular Formula C15H12N2
Beilstein 158264
Melting Point 199° to 204°C
Quantity 5g
TSCA TSCA
CAS 1145-01-3