missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Diphenyl-1-(p-tolyl)formazan, TCI America™

$155.84 - $942.66

Chemical Identifiers

CAS 13412-07-2
Molecular Formula C20H18N4
Molecular Weight (g/mol) 314.392
MDL Number MFCD00059316
InChI Key TYLDKKLAWINVKP-UHFFFAOYSA-N
Synonym p-Tolyltetrazolium Red Formazan
PubChem CID 3494325
IUPAC Name N'-anilino-N-(4-methylphenyl)iminobenzenecarboximidamide
SMILES CC1=CC=C(C=C1)N=NC(=NNC2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B0285100MG View Documents TCI America
B0285100MG
100mg Each for $155.84
 
B02851G View Documents TCI America
B02851G
1g Each for $942.66
 
Specifications

Chemical Identifiers

13412-07-2
314.392
TYLDKKLAWINVKP-UHFFFAOYSA-N
3494325
CC1=CC=C(C=C1)N=NC(=NNC2=CC=CC=C2)C3=CC=CC=C3
C20H18N4
MFCD00059316
p-Tolyltetrazolium Red Formazan
N'-anilino-N-(4-methylphenyl)iminobenzenecarboximidamide

Specifications

Undesignated
100mg
C20H18N4
p-Tolyltetrazolium Red Formazan
CC1=CC=C(C=C1)N=NC(=NNC2=CC=CC=C2)C3=CC=CC=C3
314.392
314.39
≥96.0% (HPLC)
154°C
13412-07-2
MFCD00059316
TYLDKKLAWINVKP-UHFFFAOYSA-N
N'-anilino-N-(4-methylphenyl)iminobenzenecarboximidamide
3494325
Crystal-Powder at 20°C
3,5-Diphenyl-1-(p-tolyl)formazan
Safety and Handling

Safety and Handling

TSCA : No