missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Diphenyl-1-(1-naphthyl)formazan, TCI America™

$56.53 - $215.42

Chemical Identifiers

CAS 1719-72-8
Molecular Formula C23H18N4
Molecular Weight (g/mol) 350.425
MDL Number MFCD00059534
InChI Key AXEBHHNKLXVOET-GYXHLFRYSA-N
Synonym Tetrazolium Violet Formazan, Violet Tetrazolium Formazan
PubChem CID 7064991
IUPAC Name N'-anilino-N-naphthalen-1-yliminobenzenecarboximidamide
SMILES C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC4=CC=CC=C43
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B02811G View Documents TCI America
B02811G
1g Each for $215.42
 
B0281100MG View Documents TCI America
B0281100MG
100mg Each for $56.53
 
Specifications

Chemical Identifiers

1719-72-8
350.425
AXEBHHNKLXVOET-GYXHLFRYSA-N
7064991
C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC4=CC=CC=C43
C23H18N4
MFCD00059534
Tetrazolium Violet Formazan, Violet Tetrazolium Formazan
N'-anilino-N-naphthalen-1-yliminobenzenecarboximidamide

Specifications

Undesignated
1719-72-8
MFCD00059534
AXEBHHNKLXVOET-GYXHLFRYSA-N
N'-anilino-N-naphthalen-1-yliminobenzenecarboximidamide
7064991
Crystal-Powder at 20°C
3,5-Diphenyl-1-(1-naphthyl)formazan
1g
C23H18N4
Tetrazolium Violet Formazan, Violet Tetrazolium Formazan
C1=CC=C(C=C1)C(=NNC2=CC=CC=C2)N=NC3=CC=CC4=CC=CC=C43
350.425
350.43
≥97.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No