Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dinitrosalicylaldehyde, 98%

$61.26 - $220.12

Chemical Identifiers

Chemical Name or Material 3,5-Dinitrosalicylaldehyde
CAS 2460-59-5
Molecular Formula C7H4N2O6
Formula Weight 212.12
MDL Number MFCD00007103
Beilstein 1121900
Synonym 3,5-Dinitro-2-hydroxybenzaldehyde; 2-Hydroxy-3,5-dinitrobenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2042403

 
alfa aesar™
B2042403
1g Each for $61.26

AAB2042406

 
alfa aesar™
B2042406
5g Each for $220.12
Description & Specifications

Specifications

Formula Weight 212.12
Chemical Name or Material 3,5-Dinitrosalicylaldehyde
TSCA TSCA
Assay 98%
Molecular Formula C7H4N2O6
MDL Number MFCD00007103
CAS 2460-59-5
Beilstein 1121900
Synonym 3,5-Dinitro-2-hydroxybenzaldehyde; 2-Hydroxy-3,5-dinitrobenzaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 71° to 74°C
Quantity 1g

Applications
It is used to produce 3-(2-hydroxy-3,5-dinitro-phenyl)-propenal with acetaldehyde. 3,5-Dinitrosalicylaldehyde has been used in the preparation of salicyldimine ligand via Schiff base condensation with allyl-substituted aniline, 3-hydroxycoumarins2, chromogenic proteinase substrates and ruthenium(II) chiral Schiff base complexes.

Solubility
Soluble in water, 0.6 g/100 mL (18°C).

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.