Please login to your online account to display your discounted pricing

3,5-Dinitrobenzoyl Chloride, 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC408715000

 View more versions of this product

Catalog No. AC408715000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (RCOCL)
Physical Form Crystals or Crystalline Needles
Formula Weight 230.56
Quantity 500g
Color Yellow to beige
Melting Point 67° to 71°C
Beilstein 09, 414
Fieser 01,320
Merck Index 15, 3303
CAS Max % 100.0
Molecular Formula C7H3ClN2O5
CAS 99-33-2
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Contact with water liberates toxic gas.
Chemical Name or Material 3, 5-Dinitrobenzoyl chloride
Assay Percent Range 99%
MDL Number MFCD00007248
CAS Min % 98.5
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 5-Dinitrobenzoyl chloride
CAS 99-33-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (RCOCL)
Assay Percent Range 99%
Molecular Formula C7H3ClN2O5
Formula Weight 230.56
MDL Number MFCD00007248
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Contact with water liberates toxic gas.

SDS & Safety Documents