missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dinitrobenzoyl Chloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC408710250

 View more versions of this product

Catalog No. AC408710250


 

Specifications Description & Specifications

Specifications

≥98.5% (RCOCL)
230.56
Yellow to beige
15, 3303
67° to 71°C
MFCD00007248
98.5
Authentic
99-33-2
Crystals or Crystalline Needles
25g
01,320
09, 414
100.0
3, 5-Dinitrobenzoyl chloride
Glass bottle
C7H3ClN2O5
99%

Chemical Identifiers

99-33-2
100.0
C7H3ClN2O5
MFCD00007248
≥98.5% (RCOCL)
98.5
99%
230.56
3, 5-Dinitrobenzoyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger