Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Dinitrobenzotrifluoride, 98%

$218.49 - $218.49

Chemical Identifiers

Chemical Name or Material 3,5-Dinitrobenzotrifluoride
CAS 401-99-0
Molecular Formula C7H3F3N2O4
Formula Weight 236.11
MDL Number MFCD00007233
Beilstein 2058079
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2424814

  SDS
alfa aesar™
B2424814
25g Each for $218.49
Description & Specifications

Specifications

Formula Weight 236.11
MDL Number MFCD00007233
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Chemical Name or Material 3,5-Dinitrobenzotrifluoride
Beilstein 2058079
Quantity 25g
Health Hazard 1 Warning
Melting Point 50° to 52°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H3F3N2O4
CAS 401-99-0
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.